missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl dimethylmalonate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 162660100

 View more versions of this product

Catalog No. AC162660100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Diethyl dimethylmalonate
0.9900g/mL
71°C
97%
C9H16O4
1.4114 to 1.4134
10g
0.99
UELKSYXXNGHTSE-UHFFFAOYSA-N
diethyl 2,2-dimethylpropanedioate
15382
97%
1619-62-1
192°C
Authentic
Glass bottle
C2H5O2CC(CH3)2CO2C2H5
MFCD00009128
02, II, 572
diethyl 2,2-dimethylmalonate, diethyl dimethylmalonate, propanedioic acid, dimethyl-, diethyl ester, diethyldimethylmalonate, diethyl dimethyl malonate, 1,3-diethyl 2,2-dimethylpropanedioate, malonic acid, dimethyl-, diethyl ester, 2,2-dimethyl-malonic acid diethyl ester, zlchem 328, pubchem16321
CCOC(=O)C(C)(C)C(=O)OCC
188.22
188.22

Chemical Identifiers

1619-62-1
188.22
UELKSYXXNGHTSE-UHFFFAOYSA-N
15382
CCOC(=O)C(C)(C)C(=O)OCC
C9H16O4
MFCD00009128
diethyl 2,2-dimethylmalonate, diethyl dimethylmalonate, propanedioic acid, dimethyl-, diethyl ester, diethyldimethylmalonate, diethyl dimethyl malonate, 1,3-diethyl 2,2-dimethylpropanedioate, malonic acid, dimethyl-, diethyl ester, 2,2-dimethyl-malonic acid diethyl ester, zlchem 328, pubchem16321
diethyl 2,2-dimethylpropanedioate
Safety and Handling

Safety and Handling

EINECSNumber : 216-582-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only