Learn More
Diethyl cyanophosphonate, 93%, ACROS Organics™
$222.37 - $222.37
Chemical Identifiers
CAS | 2942-58-7 |
---|---|
Molecular Formula | C5H10NO3P |
Molecular Weight (g/mol) | 163.11 |
MDL Number | MFCD00010256 |
InChI Key | ZWWWLCMDTZFSOO-UHFFFAOYSA-N |
Synonym | cyanophosphonic acid diethyl ester, diethoxy-phosphoryl cyanide, diethyl cyanidophosphate, diethyl cyanophosphonate, diethyl cyanophosphosphonate, diethyl phosphorocyanidate, diethylcyanophosphate, diethylcyanophosphonate, diethylphosphorocyanidate, phosphorocyanidic acid, diethyl ester |
PubChem CID | 76257 |
IUPAC Name | diethoxyphosphorylformonitrile |
SMILES | CCOP(=O)(C#N)OCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC224290050
|
Thermo Fisher Scientific
224290050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC224290250
|
Thermo Fisher Scientific
224290250 |
25g | Glass bottle |
Each for $222.37
|
|
AC224291000
|
Thermo Fisher Scientific
AC224291000 |
100g | Glass bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
for C-C Bond Formation (Olefination)Chemical Identifiers
2942-58-7 | |
163.11 | |
ZWWWLCMDTZFSOO-UHFFFAOYSA-N | |
76257 | |
CCOP(=O)(C#N)OCC |
C5H10NO3P | |
MFCD00010256 | |
cyanophosphonic acid diethyl ester, diethoxy-phosphoryl cyanide, diethyl cyanidophosphate, diethyl cyanophosphonate, diethyl cyanophosphosphonate, diethyl phosphorocyanidate, diethylcyanophosphate, diethylcyanophosphonate, diethylphosphorocyanidate, phosphorocyanidic acid, diethyl ester | |
diethoxyphosphorylformonitrile |
Specifications
1.0700g/mL | |
Authentic | |
1.4020 to 1.4040 | |
104.0°C to 105.0°C (19.0 mmHg) | |
5g | |
91% min. (GC) | |
(C2H5O)2P(O)CN | |
05,17; 06,192; 07,107; 10,145; 15,26; 16,24 | |
Solubility in water: decomposes in water | |
CCOP(=O)(C#N)OCC | |
163.11 | |
163.11 | |
93% |
80°C | |
Glass bottle | |
1.07 | |
Colorless to Yellow | |
2942-58-7 | |
C5H10NO3P | |
MFCD00010256 | |
cyanophosphonic acid diethyl ester, diethoxy-phosphoryl cyanide, diethyl cyanidophosphate, diethyl cyanophosphonate, diethyl cyanophosphosphonate, diethyl phosphorocyanidate, diethylcyanophosphate, diethylcyanophosphonate, diethylphosphorocyanidate, phosphorocyanidic acid, diethyl ester | |
ZWWWLCMDTZFSOO-UHFFFAOYSA-N | |
diethoxyphosphorylformonitrile | |
76257 | |
Liquid | |
Diethyl cyanophosphonate |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.
Contact with acids liberates very toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 220-936-5