missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylecyanophosphonate, 93%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 224290050

 View more versions of this product

Catalog No. AC224290050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Diethylecyanophosphonate
Authentic
1.4020 to 1.4040 (20°C, 589nm)
Clear colorless to yellow
≥91%
5g
(C2H5O)2P(O)CN
MFCD00010256
2942-58-7
Glass bottle
1.07g/mL
Liquid
93%
C5H10NO3P
163.11
Diethylphosphoryl cyanide

Chemical Identifiers

93%
(C2H5O)2P(O)CN
MFCD00010256
Diethylecyanophosphonate
C5H10NO3P
163.11
Diethylphosphoryl cyanide
≥91%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.
Contact with acids liberates very toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger