missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl carbonate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC114220100

 View more versions of this product

Catalog No. AC114220100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Alkylation reagent
Specifications

Specifications

Diethyl carbonate
33°C
Plastic drum
0.975
0.2% max. (K.F.)
Undesignated
10L
98.5% min. (GC)
(C2H5O)2CO
03,5
15,3835
Solubility in water: 18.8g/L
CCOC(=O)OCC
118.13
118.13
99%
0.9750g/mL
Authentic
1.3830 to 1.3850
0.827 mPa.s (20°C)
126.0°C to 128.0°C
-43.0°C
105-58-8
C5H10O3
MFCD00009107
01,247; 02,129; 03,95; 05,213
carbonic acid diethyl ester, carbonic acid, diethyl ester, carbonic ether, diaethylcarbonat, diatol, diethylkarbonat, ethoxyformic anhydride, ethyl carbonate, ethyl carbonate eto 2co, eufin
OIFBSDVPJOWBCH-UHFFFAOYSA-N
diethyl carbonate
7766
Liquid

Chemical Identifiers

105-58-8
118.13
OIFBSDVPJOWBCH-UHFFFAOYSA-N
7766
CCOC(=O)OCC
C5H10O3
MFCD00009107
carbonic acid diethyl ester, carbonic acid, diethyl ester, carbonic ether, diaethylcarbonat, diatol, diethylkarbonat, ethoxyformic anhydride, ethyl carbonate, ethyl carbonate eto 2co, eufin
diethyl carbonate
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-311-1

SDS
Documents
Product Certifications