missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl bis(hydroxymethyl)malonate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 189535000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Diethyl bis(hydroxymethyl)malonate | |
>110°C | |
95% | |
C9H16O6 | |
MFCD00009130 | |
diethyl bis hydroxymethyl malonate, diethyl 2,2-bis hydroxymethyl malonate, propanedioic acid, bis hydroxymethyl-, diethyl ester, 1,3-diethyl 2,2-bis hydroxymethyl propanedioate, diethyl 2,2-bis hydroxymethyl propanedioate, 2,2-bis hydroxymethyl malonic acid diethyl ester, bis hydroxymethyl malonic acid diethyl ester, diethyl 2,2-bis hydroxymethyl propane-1,3-dioate, acmc-20al8x, ksc490a8h | |
CCOC(=O)C(CO)(CO)C(=O)OCC | |
220.22 | |
220.22 |
20605-01-0 | |
Authentic | |
Glass Bottle | |
(HOCH2)2C(CO2C2H5)2 | |
500g | |
WIOHBOKEUIHYIC-UHFFFAOYSA-N | |
diethyl 2,2-bis(hydroxymethyl)propanedioate | |
311844 | |
95% |
Chemical Identifiers
20605-01-0 | |
220.22 | |
WIOHBOKEUIHYIC-UHFFFAOYSA-N | |
311844 | |
CCOC(=O)C(CO)(CO)C(=O)OCC |
C9H16O6 | |
MFCD00009130 | |
diethyl bis hydroxymethyl malonate, diethyl 2,2-bis hydroxymethyl malonate, propanedioic acid, bis hydroxymethyl-, diethyl ester, 1,3-diethyl 2,2-bis hydroxymethyl propanedioate, diethyl 2,2-bis hydroxymethyl propanedioate, 2,2-bis hydroxymethyl malonic acid diethyl ester, bis hydroxymethyl malonic acid diethyl ester, diethyl 2,2-bis hydroxymethyl propane-1,3-dioate, acmc-20al8x, ksc490a8h | |
diethyl 2,2-bis(hydroxymethyl)propanedioate |
Safety and Handling
GHS H Statement
Causes skin irritation.
GHS Signal Word: Warning
EINECSNumber : 243-916-8
RUO â Research Use Only