Please login to your online account to display your discounted pricing

Diethyldithiocarbamic Acid, Silver Salt, Reagent ACS, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  419330100

 View more versions of this product

Catalog No. AC419330100

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Amorphous Powder
Formula Weight 256.13
Quantity 10g
Color Khaki
EINECS Number 216-003-7
TSCA TSCA
Solubility Solubility in water: insoluble
Packaging Glass bottle
Suitability (for determination of arsenic) passes
Synonym Silver diethyldithiocarbamate
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material Diethyldithiocarbamic acid, silver salt
Molecular Formula C5H10AgNS2
Linear Formula (C2H5)2NCS2Ag
MDL Number MFCD00004929
CAS 1470-61-7
Health Hazard 1 Warning

Specifications

Chemical Name or Material Diethyldithiocarbamic acid, silver salt
CAS 1470-61-7
Purity Grade ACS Reagent Grade
Molecular Formula C5H10AgNS2
Linear Formula (C2H5)2NCS2Ag
Formula Weight 256.13
MDL Number MFCD00004929
Synonym Silver diethyldithiocarbamate
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents