missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylcyanamide, +99%, ACROS Organics™

Manufacturer:  Acros Organics AC200220500

 View more versions of this product

Catalog No. AC200220500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Diethylcyanamide
Authentic
1.4210 to 1.4240 (20°C, 589nm)
50g
99+%
(C2H5)2NCN
04, 121
ZZTSQZQUWBFTAT-UHFFFAOYSA-N
diethylcyanamide
12050
Fine Crystalline Powder
0.8460g/mL
Glass bottle
White
617-83-4
C5H10N2
MFCD00001769
n,n-diethylcyanamide, cyanamide, diethyl, n-cyanodiethylamine, diethylkyanamid, usaf ek-6326, diethylkyanamid czech, cyanodiethylamine, diethylamino carbonitrile, diethyl-cyanamide, aminodiethylcarbonitrile
CCN(CC)C#N
98.149
98.15
≥99.0% (GC)

Chemical Identifiers

617-83-4
98.149
ZZTSQZQUWBFTAT-UHFFFAOYSA-N
12050
CCN(CC)C#N
C5H10N2
MFCD00001769
n,n-diethylcyanamide, cyanamide, diethyl, n-cyanodiethylamine, diethylkyanamid, usaf ek-6326, diethylkyanamid czech, cyanodiethylamine, diethylamino carbonitrile, diethyl-cyanamide, aminodiethylcarbonitrile
diethylcyanamide
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents