Please login to your online account to display your discounted pricing

Diethylcyanamide, +99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Diethylcyanamide
Assay ≥99.0% (GC)
Assay Percent Range 99+%
Molecular Formula C5H10N2
Linear Formula (C2H5)2NCN
Formula Weight 98.15
MDL Number MFCD00001769
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC200220500

  SDS
thermo fisher scientific
AC200220500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC200220100

  SDS
thermo fisher scientific
AC200220100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0% (GC)
Physical Form Fine Crystalline Powder
Formula Weight 98.15
Quantity 50g
Color White
Beilstein 04, 121
Density 0.8460g/mL
Chemical Name or Material Diethylcyanamide
Assay Percent Range 99+%
Molecular Formula C5H10N2
Linear Formula (C2H5)2NCN
MDL Number MFCD00001769
CAS 617-83-4
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.4210 to 1.4240 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.