Please login to your online account to display your discounted pricing

Diethylcarbamyl Chloride 99%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  114205000

 View more versions of this product

Catalog No. AC114205000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 135.59
Assay ≥98.5% (GC)
Quantity 500mL
Color Clear
Beilstein 04, 120
TSCA TSCA
RTECS Number FD4025000
EINECS Number 201-798-5
Density 1.0700g/mL
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym Diethylcarbamoyl chloride
Refractive Index 1.4495 to 1.4525 (20°C, 589nm)
MDL Number MFCD00000636
Health Hazard 2 GHS P Statement
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Suspected of causing cancer.
Chemical Name or Material Diethylcarbamyl chloride
Health Hazard 1 Danger
Linear Formula (C2H5)2NCOCl
Assay Percent Range 99%
Name Note 99%
Molecular Formula C5H10ClNO
CAS 88-10-8

Specifications

Chemical Name or Material Diethylcarbamyl chloride
Name Note 99%
CAS 88-10-8
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C5H10ClNO
Linear Formula (C2H5)2NCOCl
Formula Weight 135.59
MDL Number MFCD00000636
Synonym Diethylcarbamoyl chloride
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Suspected of causing cancer.

SDS & Safety Documents