missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylcarbamyl Chloride 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 114201000

 View more versions of this product

Catalog No. AC114201000



Description

Description

Specifications

Specifications

Diethylcarbamyl chloride
80°C
Glass bottle
1.07
Undesignated
100mL
88-10-8
C5H10ClNO
MFCD00000636
carbamic chloride, diethyl, carbamidoyl chloride, diethyl, carbamoyl chloride, diethyl, diethylcarbamic chloride, diethylcarbamoyl chloride, diethylcarbamyl chloride, diethylchloroformamide, n,n-diethylcarbamyl chloride, n,n-diethylchloroformamide, unii-5riv1u6h5w
CCN(CC)C(=O)Cl
135.591
135.59
99%
1.0700g/mL
Authentic
1.4505 to 1.4525
117.0°C to 123.0°C (132.0 mmHg)
-32.0°C
99%
99%
(C2H5)2NCOCl
04, 120
OFCCYDUUBNUJIB-UHFFFAOYSA-N
N,N-diethylcarbamoyl chloride
6916
Liquid

Chemical Identifiers

88-10-8
135.591
OFCCYDUUBNUJIB-UHFFFAOYSA-N
6916
CCN(CC)C(=O)Cl
C5H10ClNO
MFCD00000636
carbamic chloride, diethyl, carbamidoyl chloride, diethyl, carbamoyl chloride, diethyl, diethylcarbamic chloride, diethylcarbamoyl chloride, diethylcarbamyl chloride, diethylchloroformamide, n,n-diethylcarbamyl chloride, n,n-diethylchloroformamide, unii-5riv1u6h5w
N,N-diethylcarbamoyl chloride
Safety and Handling

Safety and Handling

GHS P Statement
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Suspected of causing cancer.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 201-798-5

RTECSNumber : FD4025000

TSCA : TSCA

SDS Documents