Learn More
Diethylcarbamyl Chloride 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 114200050
Specifications
Diethylcarbamyl chloride | |
80°C | |
Glass Bottle | |
1.07 | |
Undesignated | |
5mL | |
88-10-8 | |
C5H10ClNO | |
MFCD00000636 | |
carbamic chloride, diethyl, carbamidoyl chloride, diethyl, carbamoyl chloride, diethyl, diethylcarbamic chloride, diethylcarbamoyl chloride, diethylcarbamyl chloride, diethylchloroformamide, n,n-diethylcarbamyl chloride, n,n-diethylchloroformamide, unii-5riv1u6h5w | |
CCN(CC)C(=O)Cl | |
135.591 | |
135.59 | |
99% |
1.0700g/mL | |
Authentic | |
1.4505 to 1.4525 | |
117.0°C to 123.0°C (132.0 mmHg) | |
-32.0°C | |
99% | |
99% | |
(C2H5)2NCOCl | |
04, 120 | |
OFCCYDUUBNUJIB-UHFFFAOYSA-N | |
N,N-diethylcarbamoyl chloride | |
6916 | |
Liquid |
Chemical Identifiers
88-10-8 | |
135.591 | |
OFCCYDUUBNUJIB-UHFFFAOYSA-N | |
6916 | |
CCN(CC)C(=O)Cl |
C5H10ClNO | |
MFCD00000636 | |
carbamic chloride, diethyl, carbamidoyl chloride, diethyl, carbamoyl chloride, diethyl, diethylcarbamic chloride, diethylcarbamoyl chloride, diethylcarbamyl chloride, diethylchloroformamide, n,n-diethylcarbamyl chloride, n,n-diethylchloroformamide, unii-5riv1u6h5w | |
N,N-diethylcarbamoyl chloride |
Safety and Handling
GHS P Statement
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Suspected of causing cancer.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 201-798-5
RTECSNumber : FD4025000
TSCA : TSCA