Please login to your online account to display your discounted pricing

Diethylcarbamyl Chloride 99%, ACROS Organics™

$47.79 - $158.19

Chemical Identifiers

Chemical Name or Material Diethylcarbamyl chloride
Name Note 99%
CAS 88-10-8
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C5H10ClNO
Linear Formula (C2H5)2NCOCl
Formula Weight 135.59
MDL Number MFCD00000636
Synonym Diethylcarbamoyl chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC114200050

  SDS
thermo fisher scientific
114200050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC114201000

  SDS
thermo fisher scientific
114201000
Glass bottle 100mL Each for $47.79

AC114205000

  SDS
thermo fisher scientific
114205000
Glass bottle 500mL Each for $158.19
Description & Specifications

Specifications

Formula Weight 135.59
Physical Form Liquid
Assay ≥98.5% (GC)
Quantity 5mL
Color Clear
TSCA TSCA
RTECS Number FD4025000
EINECS Number 201-798-5
Density 1.0700g/mL
Beilstein 04, 120
Refractive Index 1.4495 to 1.4525 (20°C, 589nm)
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS P Statement
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Suspected of causing cancer.
Health Hazard 1 Danger
Packaging Glass bottle
Synonym Diethylcarbamoyl chloride
Infrared Spectrum Authentic
Molecular Formula C5H10ClNO
Assay Percent Range 99%
Chemical Name or Material Diethylcarbamyl chloride
Linear Formula (C2H5)2NCOCl
Name Note 99%
CAS 88-10-8
MDL Number MFCD00000636
Safety and Handling

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Suspected of causing cancer.