missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethylbenzene, Mixture of o-, m- and p-isomers, Sum Min. 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC114162500

 View more versions of this product

Catalog No. AC114162500



Specifications

Specifications

Diethylbenzene
56°C
Glass bottle
0.869
180.0°C to 182.0°C
-73.0°C
25340-17-4
MFCD00792900
Solubility in water: insoluble. Other solubilities: soluble in alcohol, benzene, ether and carbon, tetrachloride
134.22
95%
0.8690g/mL
Authentic
1.4940 to 1.4960
0.89 mPa.s (20°C)
Colorless to Yellow
250mL
C10H14
05,IV,1067
134.22
Liquid

Chemical Identifiers

25340-17-4
C10H14
134.22
MFCD00792900
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.<b

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do

GHS Signal Word: Danger

EINECSNumber : 246-874-9

RTECSNumber : CZ5600000

TSCA : TSCA

SDS Documents