missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethylammonium diethyldithiocarbamate, 98%

$59.82 - $507.35

Chemical Identifiers

CAS 1518-58-7
Molecular Formula C9H22N2S2
MDL Number MFCD00013146
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1045814 View Documents Alfa Aesar™
A1045814
25g Each for $174.26
 
AAA1045806 View Documents Alfa Aesar™
A1045806
5g Each for $59.82
 
AAA1045822 View Documents Alfa Aesar™
A1045822
100g Each for $507.35
 
Description

Description

Applications
Diethylammonium diethyldithiocarbamate is used in the synthesis of Dithiocarbamate. It serve as hosts for the specific binding of various metal cations.

Solubility
Soluble in Chloroform, Methanol.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Incompatible with strong acids, Strong bases.
Specifications

Chemical Identifiers

1518-58-7
C9H22N2S2
MFCD00013146

Specifications

81°C to 84°C
1518-58-7
(CH3CH2)2NH2+ (CH3CH2)2NCS2-
222.42
Diethylammonium diethyldithiocarbamate
25g
C9H22N2S2
MFCD00013146
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : EZ5425000

TSCA : TSCA