Please login to your online account to display your discounted pricing

Diethylaminoacetonitrile 95%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  113991000

 View more versions of this product

Catalog No. AC113991000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥94%
Formula Weight 112.17
Quantity 100mL
Color Clear orange-brown
Beilstein 04, 350
Fieser 09,159
EINECS Number 221-130-6
TSCA TSCA
Density 0.8660g/mL
RTECS Number AL8575000
Packaging Glass bottle
Refractive Index 1.4250 to 1.4270 (20°C, 589nm)
CAS 3010-02-4
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Toxic if swallowed.
Fatal if inhaled.
Toxic in contact with skin.
Flammable liquid and vapor.
Chemical Name or Material Diethylaminoacetonitrile
Name Note 95%
Assay Percent Range 95%
Molecular Formula C6H12N2
Linear Formula (CH3CH2)2NCH2CN
MDL Number MFCD00001891
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Diethylaminoacetonitrile
Name Note 95%
CAS 3010-02-4
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H12N2
Linear Formula (CH3CH2)2NCH2CN
Formula Weight 112.17
MDL Number MFCD00001891
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Toxic if swallowed.
Fatal if inhaled.
Toxic in contact with skin.
Flammable liquid and vapor.

SDS & Safety Documents