Please login to your online account to display your discounted pricing

Diethylaminoacetonitrile 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Diethylaminoacetonitrile
Name Note 95%
CAS 3010-02-4
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H12N2
Linear Formula (CH3CH2)2NCH2CN
Formula Weight 112.17
MDL Number MFCD00001891
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC113991000

  SDS
thermo fisher scientific
113991000
Glass bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥94%
Formula Weight 112.17
Quantity 100mL
Color Clear orange-brown
Beilstein 04, 350
Fieser 09,159
TSCA TSCA
EINECS Number 221-130-6
RTECS Number AL8575000
Density 0.8660g/mL
Packaging Glass bottle
Refractive Index 1.4250 to 1.4270 (20°C, 589nm)
Infrared Spectrum Authentic
CAS 3010-02-4
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Toxic if swallowed.
Fatal if inhaled.
Toxic in contact with skin.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00001891
Assay Percent Range 95%
Chemical Name or Material Diethylaminoacetonitrile
Name Note 95%
Linear Formula (CH3CH2)2NCH2CN
Molecular Formula C6H12N2