Learn More
Diethylaminoacetaldehyde Diethyl Acetal 99%, ACROS Organics™
Chemical Identifiers
CAS | 3616-57-7 |
---|---|
Molecular Formula | C10H23NO2 |
Molecular Weight (g/mol) | 189.299 |
MDL Number | MFCD00009234 |
InChI Key | HNFUIXOBWCFVAJ-UHFFFAOYSA-N |
Synonym | 2,2-diethoxyethyl diethyl amine, 2,2-diethoxyethyl diethylamine, 2,2-diethoxytriethylamine, diethylamino acetaldehyde diethyl acetal, diethylaminoacetaldehyde diethyl acetal, diethylaminoacetaldehyde diethylacetal, ethanamine, 2,2-diethoxy-n,n-diethyl |
PubChem CID | 77164 |
IUPAC Name | 2,2-diethoxy-N,N-diethylethanamine |
SMILES | CCN(CC)CC(OCC)OCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC113971000
|
Thermo Fisher Scientific
113971000 |
100mL | Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
3616-57-7 | |
189.299 | |
HNFUIXOBWCFVAJ-UHFFFAOYSA-N | |
77164 | |
CCN(CC)CC(OCC)OCC |
C10H23NO2 | |
MFCD00009234 | |
2,2-diethoxyethyl diethyl amine, 2,2-diethoxyethyl diethylamine, 2,2-diethoxytriethylamine, diethylamino acetaldehyde diethyl acetal, diethylaminoacetaldehyde diethyl acetal, diethylaminoacetaldehyde diethylacetal, ethanamine, 2,2-diethoxy-n,n-diethyl | |
2,2-diethoxy-N,N-diethylethanamine |
Specifications
0.8500g/mL | |
Glass Bottle | |
Yellow to Brown | |
3616-57-7 | |
C10H23NO2 | |
MFCD00009234 | |
HNFUIXOBWCFVAJ-UHFFFAOYSA-N | |
2,2-diethoxy-N,N-diethylethanamine | |
77164 | |
Liquid | |
Diethylaminoacetaldehyde diethyl acetal, 99% |
Authentic | |
1.4189 to 1.4209 (20°C, 589nm) | |
100mL | |
99% | |
(C2H5)2NCH2CH(OC2H5)2 | |
2,2-diethoxyethyl diethyl amine, 2,2-diethoxyethyl diethylamine, 2,2-diethoxytriethylamine, diethylamino acetaldehyde diethyl acetal, diethylaminoacetaldehyde diethyl acetal, diethylaminoacetaldehyde diethylacetal, ethanamine, 2,2-diethoxy-n,n-diethyl | |
CCN(CC)CC(OCC)OCC | |
189.299 | |
189.3 | |
≥98.5% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MOISTURE SENSITIVE
EINECSNumber : 222-801-6