Please login to your online account to display your discounted pricing

Diethylamine (Reagent), Fisher Chemical


Quantity 1L
Packaging Glass Bottle

Manufacturer: fisher scientific company  D461

 View more versions of this product

Catalog No. D46-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 55°C
Quantity 1L
Formula Weight 73.14
Melting Point −50°C
Purity Grade Reagent Grade
Flash Point −23°C
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
pH 12.0
Molecular Formula C4H11N
Color Colorless
Density 0.710g/cm³
UN Number 1154
Identification Pass Test
MDL Number MFCD00009032
Cleanup Symbol Solvent
Packaging Glass Bottle
Synonym N-Ethylethanamine
Boiling Range 3°C max.
ChemAlert Storage Symbol Red
Linear Formula (C2H5)2NH
Chemical Name or Material Diethylamine
CAS 109-89-7

Specifications

Chemical Name or Material Diethylamine
CAS 109-89-7
Purity Grade Reagent Grade
Molecular Formula C4H11N
Linear Formula (C2H5)2NH
Formula Weight 73.14
MDL Number MFCD00009032
Synonym N-Ethylethanamine
Accessories
   

SDS & Safety Documents