Learn More
Diethylamine, 99+%, Extra Pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 149450100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Diethylamine | |
0.7100g/mL | |
−23°C | |
99% min. (GC) | |
C4H11N | |
(C2H5)2NH | |
10L | |
04,148; 13,312; 14,191 | |
15, 3122 | |
Solubility in water: soluble. Other solubilities: miscible with alcohol and most common organic, solvents | |
CCNCC | |
73.14 | |
0.34 mPa.s (25°C) | |
73.13 | |
Extra Pure |
109-89-7 | |
55.0°C | |
Authentic | |
Plastic drum | |
1.3840 to 1.3860 | |
MFCD00009032 | |
04, 95 | |
0.71 | |
diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine | |
HPNMFZURTQLUMO-UHFFFAOYSA-N | |
N-ethylethanamine | |
8021 | |
CHEBI:85259 | |
99+% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 203-716-3
RUO â Research Use Only