missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethylaluminum ethoxide, 25% w/w in hexane

Manufacturer:  Alfa Aesar™ 87625NE

 View more versions of this product

Catalog No. AA87625NE



Specifications

Specifications

Diethylaluminum ethoxide
1586-92-1
−23°C (−9°F)
Colorless
Air & Moisture Sensitive
(C2H5)2Al(OC2H5)
UN3394
GCPCLEKQVMKXJM-UHFFFAOYSA-N
ethoxy(diethyl)alumane
16683109
Liquid
25% w/w in hexane
50g
0.851g/mL
Petrol-like
C6H15AlO
MFCD00009059
diethylaluminum ethoxide, aluminum, ethoxydiethyl, ethoxydiethylaluminium, diethylaluminium ethoxide, ethoxydiethylaluminum, ethoxy-diethylaluminum, ethoxydiethyl aluminum, ethoxy-diethylaluminium, ethoxydiethyl aluminium, ethoxydiethyl-aluminium
CCO[Al](CC)CC
130.167
130.17

Chemical Identifiers

1586-92-1
130.167
GCPCLEKQVMKXJM-UHFFFAOYSA-N
16683109
CCO[Al](CC)CC
C6H15AlO
MFCD00009059
diethylaluminum ethoxide, aluminum, ethoxydiethyl, ethoxydiethylaluminium, diethylaluminium ethoxide, ethoxydiethylaluminum, ethoxy-diethylaluminum, ethoxydiethyl aluminum, ethoxy-diethylaluminium, ethoxydiethyl aluminium, ethoxydiethyl-aluminium
ethoxy(diethyl)alumane
Safety and Handling

Safety and Handling

GHS H Statement
H250-H314-H361-H373-H336
Catches fire spontaneously if exposed to air.
Causes severe skin burns and eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.

GHS P Statement
P222-P280-P273-P305+P351+P338-P310-P370+P378i
Do not allow contact with air.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE

TSCA : TSCA

SDS Documents