Please login to your online account to display your discounted pricing

Diethylaluminium iodide, 0.9M solution in toluene, AcroSeal™, ACROS Organics™


Quantity 800mL

Manufacturer: thermo fisher scientific  AC381168000

 View more versions of this product

Catalog No. AC381168000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure. Suspected of damaging fertility or the unborn child. Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. Reacts violently with water.
Health Hazard 3 GHS P Statement
Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water.
Physical Form Liquid
Formula Weight 212.01
Quantity 800mL
Color Clear colorless
Linear Formula (C2H5)2AlI
CAS 108-88-3
MDL Number MFCD00001025
Molecular Formula C4H10AlI
Name Note 0.8M Solution in Toluene
Synonym DEAI
Health Hazard 1 Danger
CAS 2040-00-8
Chemical Name or Material Diethylaluminium iodide

Specifications

Chemical Name or Material Diethylaluminium iodide
Name Note 0.8M Solution in Toluene
CAS 2040-00-8
CAS 108-88-3
Molecular Formula C4H10AlI
Linear Formula (C2H5)2AlI
Formula Weight 212.01
MDL Number MFCD00001025
Synonym DEAI
Accessories

Danger

GHS P Statement
Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure. Suspected of damaging fertility or the unborn child. Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. Reacts violently with water.

SDS & Safety Documents