Please login to your online account to display your discounted pricing

Diethylaluminium chloride, 1M solution in hexanes, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  199528000

 View more versions of this product

Catalog No. AC199528000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Store in a dry place. Store in a closed container.
Physical Form Solution
Formula Weight 120.56
Quantity 800mL
Color Colorless
Packaging AcroSeal™ Glass Bottle
RTECS Number BD0558000
TSCA TSCA
EINECS Number 202-477-2
Concentration 0.95 to 1.10M
Density 0.7110g/mL
CAS 110-54-3
Synonym DEAC
Health Hazard 1 Danger
CAS 96-10-6
Molecular Formula C4H10AlCl
Chemical Name or Material Diethylaluminium chloride
Name Note 1M Solution in Hexanes
Linear Formula (C2H5)2AICl
MDL Number MFCD00000459

Specifications

Chemical Name or Material Diethylaluminium chloride
Name Note 1M Solution in Hexanes
CAS 96-10-6
CAS 110-54-3
Molecular Formula C4H10AlCl
Linear Formula (C2H5)2AICl
Formula Weight 120.56
MDL Number MFCD00000459
Synonym DEAC
Accessories

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Store in a dry place. Store in a closed container.

GHS H Statement
Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.

SDS & Safety Documents