missing translation for 'onlineSavingsMsg'
Learn More

Diethyl tetrafluorosuccinate, 97%

CAS: 377-71-9 | C8H10F4O4 | 246.158 g/mol

$52.25 - $146.68

Chemical Identifiers

CAS 377-71-9
Molecular Formula C8H10F4O4
Molecular Weight (g/mol) 246.158
MDL Number MFCD00015155
InChI Key BLZSSBPZHBFNBN-UHFFFAOYSA-N
Synonym diethyl tetrafluorosuccinate, diethyl perfluorosuccinate, butanedioic acid, tetrafluoro-, diethyl ester, diethyl 2,2,3,3-tetrafluorosuccinate, butanedioic acid, 2,2,3,3-tetrafluoro-, 1,4-diethyl ester, 1,4-diethyl 2,2,3,3-tetrafluorobutanedioate, diethyl 2,2,3,3-tetrafluorobutane-1,4-dioate, diethyltetrafluorosuccinate, ethyl perfluorosuccinate, acmc-1cqkj
PubChem CID 67834
IUPAC Name diethyl 2,2,3,3-tetrafluorobutanedioate
SMILES CCOC(=O)C(C(C(=O)OCC)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1646903
View Documents
Thermo Scientific Chemicals
L1646903
1 g
Each for $52.25
Only null left
 
AAL1646906
View Documents
Thermo Scientific Chemicals
L1646906
5 g
Each for $146.68
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

377-71-9
246.158
BLZSSBPZHBFNBN-UHFFFAOYSA-N
67834
CCOC(=O)C(C(C(=O)OCC)(F)F)(F)F
C8H10F4O4
MFCD00015155
diethyl tetrafluorosuccinate, diethyl perfluorosuccinate, butanedioic acid, tetrafluoro-, diethyl ester, diethyl 2,2,3,3-tetrafluorosuccinate, butanedioic acid, 2,2,3,3-tetrafluoro-, 1,4-diethyl ester, 1,4-diethyl 2,2,3,3-tetrafluorobutanedioate, diethyl 2,2,3,3-tetrafluorobutane-1,4-dioate, diethyltetrafluorosuccinate, ethyl perfluorosuccinate, acmc-1cqkj
diethyl 2,2,3,3-tetrafluorobutanedioate

Specifications

377-71-9
78°C (5 mmHg)
1.368
1 g
diethyl tetrafluorosuccinate, diethyl perfluorosuccinate, butanedioic acid, tetrafluoro-, diethyl ester, diethyl 2,2,3,3-tetrafluorosuccinate, butanedioic acid, 2,2,3,3-tetrafluoro-, 1,4-diethyl ester, 1,4-diethyl 2,2,3,3-tetrafluorobutanedioate, diethyl 2,2,3,3-tetrafluorobutane-1,4-dioate, diethyltetrafluorosuccinate, ethyl perfluorosuccinate, acmc-1cqkj
CCOC(=O)C(C(C(=O)OCC)(F)F)(F)F
246.158
246.16
Diethyl tetrafluorosuccinate
1.273
C8H10F4O4
MFCD00015155
1798175
BLZSSBPZHBFNBN-UHFFFAOYSA-N
diethyl 2,2,3,3-tetrafluorobutanedioate
67834
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 206-821-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only