Please login to your online account to display your discounted pricing

Diethyl Terephthalate 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  407451000

 View more versions of this product

Catalog No. AC407451000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Low Melting Solid
Formula Weight 222.24
Quantity 100g
Color White
EINECS Number 211-249-1
TSCA TSCA
RTECS Number WZ1223400
Melting Point 42° to 45°C
Density 1.11g/mL
Chemical Name or Material Diethyl terephthalate
Name Note 98%
MDL Number MFCD00039891
CAS 636-09-9
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C12H14O4

Specifications

Chemical Name or Material Diethyl terephthalate
Name Note 98%
CAS 636-09-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H14O4
Formula Weight 222.24
MDL Number MFCD00039891