missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl terephthalate, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2140118
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethyl terephthalate | |
44°C to 46°C | |
C12H14O4 | |
50 g | |
ONIHPYYWNBVMID-UHFFFAOYSA-N | |
diethyl benzene-1,4-dicarboxylate | |
12483 | |
95% |
636-09-9 | |
1.1 | |
MFCD00039891 | |
diethyl terephthalate, p-diethyl phthalate, diethylterephthalate, terephthalic acid, diethyl ester, 1,4-benzenedicarboxylic acid, diethyl ester, diethyl p-phthalate, terephthalic acid diethyl ester, unii-n97x85l3cd, 1,4-diethyl benzene-1,4-dicarboxylate, 1,4-benzenedicarboxylic acid, 1,4-diethyl ester | |
CCOC(=O)C1=CC=C(C=C1)C(=O)OCC | |
222.24 | |
222.24 |
Chemical Identifiers
636-09-9 | |
222.24 | |
ONIHPYYWNBVMID-UHFFFAOYSA-N | |
12483 | |
CCOC(=O)C1=CC=C(C=C1)C(=O)OCC |
C12H14O4 | |
MFCD00039891 | |
diethyl terephthalate, p-diethyl phthalate, diethylterephthalate, terephthalic acid, diethyl ester, 1,4-benzenedicarboxylic acid, diethyl ester, diethyl p-phthalate, terephthalic acid diethyl ester, unii-n97x85l3cd, 1,4-diethyl benzene-1,4-dicarboxylate, 1,4-benzenedicarboxylic acid, 1,4-diethyl ester | |
diethyl benzene-1,4-dicarboxylate |
Safety and Handling
EINECSNumber : 211-249-1
RTECSNumber : WZ1223400
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only