missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl sulfate, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 114560010
Description
Alkylation reagentSpecifications
Diethyl sulfate | |
104°C | |
Glass bottle | |
1.1795 | |
0.15% max. | |
Undesignated | |
1L | |
98.5% min. (GC) | |
(C2H5O)2SO2 | |
01,327 | |
15,3149 | |
Solubility in water: 5g/L (20°C). Other solubilities: decomposes in water,soluble in alcohol and ether | |
CCOS(=O)(=O)OCC | |
154.18 | |
CHEBI:34699 | |
Liquid |
1.1795g/mL | |
Authentic | |
1.3990 to 1.4010 | |
1.79 mPa.s (20°C) | |
208.0°C | |
-24.0°C | |
64-67-5 | |
C4H10O4S | |
MFCD00009099 | |
01,253 | |
des van, diaethylsulfat, diaethylsulfat german, diethyl sulphate, diethyl tetraoxosulfate, diethylester kyseliny sirove, diethylsulfate, sulfuric acid diethyl ester, sulfuric acid, diethyl ester, unii-k0fo4vfa7i | |
DENRZWYUOJLTMF-UHFFFAOYSA-N | |
diethyl sulfate | |
6163 | |
154.18 | |
99% |
Chemical Identifiers
64-67-5 | |
154.18 | |
DENRZWYUOJLTMF-UHFFFAOYSA-N | |
6163 | |
diethyl sulfate |
C4H10O4S | |
MFCD00009099 | |
des van, diaethylsulfat, diaethylsulfat german, diethyl sulphate, diethyl tetraoxosulfate, diethylester kyseliny sirove, diethylsulfate, sulfuric acid diethyl ester, sulfuric acid, diethyl ester, unii-k0fo4vfa7i | |
CHEBI:34699 | |
CCOS(=O)(=O)OCC |
Safety and Handling
GHS H Statement
May cause genetic defects.
May cause cancer.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger
EINECSNumber : 200-589-6