Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl suberate, 99%

$144.65 - $1,069.80

Chemical Identifiers

Chemical Name or Material Diethyl suberate
CAS 2050-23-9
Molecular Formula C12H22O4
Linear Formula CH3CH2O2C(CH2)6CO2CH2CH3
Formula Weight 230.3
MDL Number MFCD00009217
Beilstein 1785500
Synonym Octanedioic acid diethyl ester; Suberic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1473518

  SDS
alfa aesar™
A1473518
50g Each for $144.65

AAA1473530

  SDS
alfa aesar™
A1473530
250g Each for $583.12

AAA1473536

  SDS
alfa aesar™
A1473536
500g Each for $1,069.80
Description & Specifications

Specifications

Formula Weight 230.3
Flash Point >110°C (230°F)
Quantity 50g
Synonym Octanedioic acid diethyl ester; Suberic acid diethyl ester
Molecular Formula C12H22O4
CAS 2050-23-9
Assay 99%
MDL Number MFCD00009217
Linear Formula CH3CH2O2C(CH2)6CO2CH2CH3
Beilstein 1785500
Chemical Name or Material Diethyl suberate
Boiling Point 282° to 283°C