missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl suberate, 99%

Manufacturer:  Alfa Aesar™ A1473530

 View more versions of this product

Catalog No. AAA1473530



Specifications

Specifications

Diethyl suberate
282°C to 283°C
2050-23-9
CH3CH2O2C(CH2)6CO2CH2CH3
1785500
PEUGOJXLBSIJQS-UHFFFAOYSA-N
diethyl octanedioate
16301
99%
>110°C (230°F)
250g
C12H22O4
MFCD00009217
diethyl suberate, ethyl suberate, octanedioic acid, diethyl ester, diethylsuberate, suberic acid diethyl ester, 1,8-diethyl octanedioate, acmc-1ce5z, ksc492k9j, octanedioic acid diethyl ester, octanedioic acid,1,8-diethyl ester
CCOC(=O)CCCCCCC(=O)OCC
230.304
230.3

Chemical Identifiers

2050-23-9
230.304
PEUGOJXLBSIJQS-UHFFFAOYSA-N
16301
CCOC(=O)CCCCCCC(=O)OCC
C12H22O4
MFCD00009217
diethyl suberate, ethyl suberate, octanedioic acid, diethyl ester, diethylsuberate, suberic acid diethyl ester, 1,8-diethyl octanedioate, acmc-1ce5z, ksc492k9j, octanedioic acid diethyl ester, octanedioic acid,1,8-diethyl ester
diethyl octanedioate
SDS Documents