Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl sec-butylmalonate, 96%

$60.66 - $195.49

Chemical Identifiers

Chemical Name or Material Diethyl sec-butylmalonate
CAS 83-27-2
Molecular Formula C11H20O4
Formula Weight 216.28
MDL Number MFCD00015194
Beilstein 1709561
Synonym sec-Butylmalonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1225314

 
alfa aesar™
L1225314
25g Each for $195.49

AAL1225306

 
alfa aesar™
L1225306
5g Each for $60.66
Description & Specifications

Specifications

Formula Weight 216.28
Chemical Name or Material Diethyl sec-butylmalonate
Assay 96%
Molecular Formula C11H20O4
MDL Number MFCD00015194
CAS 83-27-2
Beilstein 1709561
Boiling Point 110° to 114°C (18mmHg)
Synonym sec-Butylmalonic acid diethyl ester
Quantity 25g