missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl sec-butylmalonate, 96%

Manufacturer:  Alfa Aesar™ L1225306

 View more versions of this product

Catalog No. AAL1225306



Specifications

Specifications

Diethyl sec-butylmalonate
5g
C11H20O4
1709561
MIIZSUOEOUHAIZ-UHFFFAOYSA-N
diethyl 2-butan-2-ylpropanedioate
78940
96%
110°C to 114°C (18mmHg)
83-27-2
MFCD00015194
diethyl sec-butylmalonate, diethyl 2-sec-butyl malonate, sec-butylmalonic acid diethyl ester, diethyl 1-methylpropyl malonate, diethyl 1-methylpropylmalonate, sec-butylmalonic acid, diethyl ester, 1,3-diethyl 2-sec-butyl propanedioate, diethyl s-butylmalonate, diethylsec-butylmalonate, acmc-209prt
CCC(C)C(C(=O)OCC)C(=O)OCC
216.277
216.28

Chemical Identifiers

83-27-2
216.277
MIIZSUOEOUHAIZ-UHFFFAOYSA-N
78940
CCC(C)C(C(=O)OCC)C(=O)OCC
C11H20O4
MFCD00015194
diethyl sec-butylmalonate, diethyl 2-sec-butyl malonate, sec-butylmalonic acid diethyl ester, diethyl 1-methylpropyl malonate, diethyl 1-methylpropylmalonate, sec-butylmalonic acid, diethyl ester, 1,3-diethyl 2-sec-butyl propanedioate, diethyl s-butylmalonate, diethylsec-butylmalonate, acmc-209prt
diethyl 2-butan-2-ylpropanedioate
Documents