missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl pyrocarbonate, 97%, pure, ACROS Organics™

Manufacturer:  Acros Organics AC170251000

 View more versions of this product

Catalog No. AC170251000



Specifications

Specifications

Diethyl pyrocarbonate
69°C
1.3960 to 1.4000
100g
97%
O(CO2C2H5)2
diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
CCOC(=O)OC(=O)OCC
162.141
CHEBI:59051
97%
1.1175g/mL
Glass bottle
93.0°C to 94.0°C (18.0mmHg)
1609-47-8
C6H10O5
MFCD00009106
FFYPMLJYZAEMQB-UHFFFAOYSA-N
ethoxycarbonyl ethyl carbonate
3051
162.14
Pure

Chemical Identifiers

1609-47-8
162.141
FFYPMLJYZAEMQB-UHFFFAOYSA-N
3051
ethoxycarbonyl ethyl carbonate
C6H10O5
MFCD00009106
diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
CHEBI:59051
CCOC(=O)OC(=O)OCC
Safety and Handling

Safety and Handling

GHS P Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

SDS Documents