missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl phthalate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1752930

 View more versions of this product

Catalog No. AAA1752930



Description

Description

Diethyl phthalate is used in the production of perfumes, acetyl salicylic acid, plastics and mosquito repellents. It is used in cosmetic formulations and personal care products such as sprays, nail polishes, and perfumes. It acts as an alcohol denaturant. It is also used as a camphor substitute, plasticizer in solid rocket propellants and wetting agent. It finds application as a surface lubricant in food and pharmaceutical packaging. In addition to this, it is used as a detergent base and aerosol spray.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl phthalate is used in the production of perfumes, acetyl salicylic acid, plastics and mosquito repellents. It is used in cosmetic formulations and personal care products such as sprays, nail polishes, and perfumes. It acts as an alcohol denaturant. It is also used as a camphor substitute, plasticizer in solid rocket propellants and wetting agent. It finds application as a surface lubricant in food and pharmaceutical packaging. In addition to this, it is used as a detergent base and aerosol spray.

Solubility
Miscible with ethanol, chloroform, diethyl ether, benzene, acetone, vegetable oils, ketones, carbon tetrachloride, esters, aromatic hydrocarbons and methanol. Slightly miscible with water and aliphatic solvents.

Notes
Incompatible with strong oxidizing agents, strong acids, permanganates and alkalies. Store in a cool place.
Specifications

Specifications

Diethyl phthalate
-64°C
296°C to 298°C
C12H14O4
MFCD00009111
1912500
diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e
FLKPEMZONWLCSK-UHFFFAOYSA-N
diethyl benzene-1,2-dicarboxylate
6781
222.24
84-66-2
1.118
160°C (320°F)
1.502
250 g
14,7371
Miscible with ethanol,chloroform,diethyl ether,benzene,acetone,vegetable oils,ketones,carbon tetrachloride,esters,aromatic hydrocarbons and methanol. Slightly miscible with water and aliphatic solvents.
CCOC(=O)C1=CC=CC=C1C(=O)OCC
222.24
CHEBI:34698
99%

Chemical Identifiers

84-66-2
222.24
FLKPEMZONWLCSK-UHFFFAOYSA-N
6781
diethyl benzene-1,2-dicarboxylate
C12H14O4
MFCD00009111
diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e
CHEBI:34698
CCOC(=O)C1=CC=CC=C1C(=O)OCC
Safety and Handling

Safety and Handling

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 201-550-6

RTECSNumber : TI1050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only