Please login to your online account to display your discounted pricing

Diethyl Phosphonoacetaldehyde Diethyl Acetal (Tech.) 90%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  114510250

 View more versions of this product

Catalog No. AC114510250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 254.26
Physical Form Liquid
Assay ≥88%
Quantity 25g
Color Clear colorless
EINECS Number 231-504-0
Density 1.0520g/mL
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4260 to 1.4320 (20°C, 589nm)
MDL Number MFCD00009236
Packaging Glass bottle
Synonym Diethyl 2, 2-diethoxyethylphosphonate
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Name Note tech. 90%
Molecular Formula C10 H23 O5 P
Chemical Name or Material Diethyl phosphonoacetaldehyde diethyl acetal
Linear Formula (C2H5O)2P(O)CH2CH(OC2H5)2
CAS 7598-61-0
Assay Percent Range 90%

Specifications

Chemical Name or Material Diethyl phosphonoacetaldehyde diethyl acetal
Name Note tech. 90%
CAS 7598-61-0
Purity Grade Technical Grade
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C10 H23 O5 P
Linear Formula (C2H5O)2P(O)CH2CH(OC2H5)2
Formula Weight 254.26
MDL Number MFCD00009236
Synonym Diethyl 2, 2-diethoxyethylphosphonate
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents