Please login to your online account to display your discounted pricing

Diethyl Phosphonoacetaldehyde Diethyl Acetal (Tech.) 90%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Diethyl phosphonoacetaldehyde diethyl acetal
Name Note tech. 90%
CAS 7598-61-0
Purity Grade Technical Grade
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C10 H23 O5 P
Linear Formula (C2H5O)2P(O)CH2CH(OC2H5)2
Formula Weight 254.26
MDL Number MFCD00009236
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC114510250

  SDS
fisher scientific acros
114510250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Technical Grade
Physical Form Liquid
Assay ≥88%
Formula Weight 254.26
Quantity 25g
Color Clear colorless
EINECS Number 231-504-0
Density 1.0520g/mL
Infrared Spectrum Authentic
Chemical Name or Material Diethyl phosphonoacetaldehyde diethyl acetal
Refractive Index 1.4260 to 1.4320 (20°C, 589nm)
Name Note tech. 90%
Molecular Formula C10 H23 O5 P
Linear Formula (C2H5O)2P(O)CH2CH(OC2H5)2
MDL Number MFCD00009236
Synonym Diethyl 2, 2-diethoxyethylphosphonate
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 90%
CAS 7598-61-0

For C-C Bond Formation (Olefination)

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.