missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl phosphite, 97+%

Manufacturer:  Alfa Aesar™ 4400014

 View more versions of this product

Catalog No. AA4400014



Description

Description

  • Packaged under Argon
Specifications

Specifications

Diethyl phosphite
74°C (165°F)
Moisture Sensitive
Colorless
25g
C4H10O3P+
diethyl phosphite, diethyl phosphonate, phosphonic acid, diethyl ester, phosphonic acid diethyl ester, diethoxyphosphine oxide, hydrogen diethyl phosphite, diethyl acid phosphite, o,o-diethyl phosphonate, phosphorous acid, diethyl ester, ethyl phosphonate eto 2hpo
CCO[P+](=O)OCC
137.095
138.1
≥97%
1.070g/mL
Weak
50°C to 51°C (2mmHg)
-70°C
762-04-9
MFCD00044573
LXCYSACZTOKNNS-UHFFFAOYSA-N
diethoxy(oxo)phosphanium
6327654
Liquid

Chemical Identifiers

762-04-9
137.095
LXCYSACZTOKNNS-UHFFFAOYSA-N
6327654
CCO[P+](=O)OCC
C4H10O3P+
MFCD00044573
diethyl phosphite, diethyl phosphonate, phosphonic acid, diethyl ester, phosphonic acid diethyl ester, diethoxyphosphine oxide, hydrogen diethyl phosphite, diethyl acid phosphite, o,o-diethyl phosphonate, phosphorous acid, diethyl ester, ethyl phosphonate eto 2hpo
diethoxy(oxo)phosphanium
Safety and Handling

Safety and Handling

GHS H Statement
H318-H317-H227
Causes serious eye damage.
May cause an allergic skin reaction.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : TG7875000

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

Documents