Learn More
Diethyl phosphite, 96%, Thermo Scientific Chemicals
Intermediate for preparation of phosphonate esters
Supplier: Thermo Scientific Chemicals 04380130
Description
Diethyl phosphite used as intermediate for extraction agent and phosphoric acid esters. It is also used as solvent and acid acceptor in the synthesis of polypeptide. It is also used in the synthetic reaction. It is a precursor of phosphorous which is used for the synthesis of various organophosphorus compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethyl phosphite | |
-70°C | |
50°C to 51°C (2 mmHg) | |
Weak | |
MFCD00044573 | |
Moisture sensitive | |
Miscible in water. | |
CCO[P+](=O)OCC | |
137.095 | |
138.1 | |
Liquid |
762-04-9 | |
1.070 g/mL | |
74°C (165°F) | |
C4H10O3P+ | |
250 g | |
diethyl phosphite, diethyl phosphonate, phosphonic acid, diethyl ester, phosphonic acid diethyl ester, diethoxyphosphine oxide, hydrogen diethyl phosphite, diethyl acid phosphite, o,o-diethyl phosphonate, phosphorous acid, diethyl ester, ethyl phosphonate eto 2hpo | |
LXCYSACZTOKNNS-UHFFFAOYSA-N | |
diethoxy(oxo)phosphanium | |
6327654 | |
96% |
Chemical Identifiers
762-04-9 | |
137.095 | |
LXCYSACZTOKNNS-UHFFFAOYSA-N | |
6327654 | |
CCO[P+](=O)OCC |
C4H10O3P+ | |
MFCD00044573 | |
diethyl phosphite, diethyl phosphonate, phosphonic acid, diethyl ester, phosphonic acid diethyl ester, diethoxyphosphine oxide, hydrogen diethyl phosphite, diethyl acid phosphite, o,o-diethyl phosphonate, phosphorous acid, diethyl ester, ethyl phosphonate eto 2hpo | |
diethoxy(oxo)phosphanium |
Safety and Handling
GHS H Statement
H318-H317-H227-H315
Causes serious eye damage.
May cause an allergic skin reaction.
Combustible liquid.
Causes skin irritation.
P210-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P370+P378q-P501c
H227-H317-H318
EINECSNumber : 212-091-6
RTECSNumber : TG7875000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only