Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl phenylthiomethylphosphonate

Chemical Identifiers

Chemical Name or Material Diethyl phenylthiomethylphosphonate
CAS 38066-16-9
Formula Weight 260.30g/mol
MDL Number MFCD00015130
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1036906

 
alfa aesar™
10369-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1036914

 
alfa aesar™
10369-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 1.174g/mL
Quantity 5g
Boiling Point 147°C (0.5mmHg)
Quantity 5g
Chemical Name or Material Diethyl phenylthiomethylphosphonate
CAS 38066-16-9
Physical Form Liquid
Formula Weight 260.30g/mol
MDL Number MFCD00015130