missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl phenylphosphonite, 98%, Thermo Scientific Chemicals
$255.67 - $3250.40
Chemical Identifiers
CAS | 1638-86-4 |
---|---|
Molecular Formula | C10H15O2P |
Molecular Weight (g/mol) | 198.20 |
MDL Number | MFCD00009086 |
InChI Key | RVDJLKVICMLVJQ-UHFFFAOYSA-N |
Synonym | diethyl phenylphosphonite, diethoxyphenylphosphine, phosphonous acid, phenyl-, diethyl ester, diethyl benzenephosphonite, diethyl phenyl-phosphonite, phosphonous acid, p-phenyl-, diethyl ester, pubchem6466, diethylphenylphosphonite, diethyl phenylphophonite, acmc-20al8o |
PubChem CID | 74225 |
IUPAC Name | diethoxy(phenyl)phosphane |
SMILES | CCOP(OCC)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2483606
|
Thermo Scientific Chemicals
B2483606 |
5 g |
Each for $255.67
|
|
|||||
AAB2483614
|
Thermo Scientific Chemicals
B2483614 |
25 g |
Each for $1,010.68
|
|
|||||
AAB2483622
|
Thermo Scientific Chemicals
B2483622 |
100 g |
Each for $3,250.40
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1638-86-4 | |
198.20 | |
RVDJLKVICMLVJQ-UHFFFAOYSA-N | |
74225 | |
CCOP(OCC)C1=CC=CC=C1 |
C10H15O2P | |
MFCD00009086 | |
diethyl phenylphosphonite, diethoxyphenylphosphine, phosphonous acid, phenyl-, diethyl ester, diethyl benzenephosphonite, diethyl phenyl-phosphonite, phosphonous acid, p-phenyl-, diethyl ester, pubchem6466, diethylphenylphosphonite, diethyl phenylphophonite, acmc-20al8o | |
diethoxy(phenyl)phosphane |
Specifications
1638-86-4 | |
64°C (0.7 mmHg) | |
C10H15O2P | |
MFCD00009086 | |
2804900 | |
diethyl phenylphosphonite, diethoxyphenylphosphine, phosphonous acid, phenyl-, diethyl ester, diethyl benzenephosphonite, diethyl phenyl-phosphonite, phosphonous acid, p-phenyl-, diethyl ester, pubchem6466, diethylphenylphosphonite, diethyl phenylphophonite, acmc-20al8o | |
CCOP(OCC)C1=CC=CC=C1 | |
198.20 | |
198.2 | |
Diethyl phenylphosphonite |
1.032 | |
>110°C (230°F) | |
1.51 | |
5 g | |
Air sensitive | |
RVDJLKVICMLVJQ-UHFFFAOYSA-N | |
diethoxy(phenyl)phosphane | |
74225 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 216-676-7
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only