Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl phenylphosphonate, 97%

Chemical Identifiers

Chemical Name or Material Diethyl phenylphosphonate
CAS 1754-49-0
Molecular Formula C10H15O3P
Formula Weight 214.20g/mol
MDL Number MFCD00015136
Beilstein 2449931
Synonym Phenylphosphonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0924006

 
alfa aesar™
L09240-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 267°C
Flash Point >110°C (230°F)
Quantity 5g
Chemical Name or Material Diethyl phenylphosphonate
CAS 1754-49-0
Assay 97%
Molecular Formula C10H15O3P
Formula Weight 214.20g/mol
MDL Number MFCD00015136
Beilstein 2449931
Synonym Phenylphosphonic acid diethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number TA0377000
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.