missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl Phenyl Orthoformate 97.0+%, TCI America™

Manufacturer:  TCI America O01875G

 View more versions of this product

Catalog No. O01875G



Specifications

Specifications

Diethyl Phenyl Orthoformate
Colorless
5g
C11H16O3
acmc-209csr
CCOC(OCC)OC1=CC=CC=C1
196.246
196.25
≥97.0% (GC)
104°C
-26°C
14444-77-0
MFCD00009228
QTURWMMVIIBRRP-UHFFFAOYSA-N
diethoxymethoxybenzene
84440
Clear Liquid at 20°C

Chemical Identifiers

14444-77-0
196.246
QTURWMMVIIBRRP-UHFFFAOYSA-N
84440
CCOC(OCC)OC1=CC=CC=C1
C11H16O3
MFCD00009228
acmc-209csr
diethoxymethoxybenzene
Safety and Handling

Safety and Handling

TSCA : No

Documents