missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl Oxalacetate 95.0+%, TCI America™
Supplier: TCI America O007325G
Specifications
Diethyl Oxalacetate | |
Yellow | |
C8H12O5 | |
25 g | |
JDXYSCUOABNLIR-UHFFFAOYSA-N | |
diethyl 2-oxobutanedioate | |
66951 | |
188.18 | |
Liquid |
108-56-5 | |
87°C | |
MFCD00059229 | |
Ethyl Oxalacetate, Oxalacetic Acid Diethyl Ester | |
CCOC(=O)CC(=O)C(=O)OCC | |
188.179 | |
CHEBI:86608 | |
≥95.0% (GC) |
Chemical Identifiers
108-56-5 | |
188.179 | |
JDXYSCUOABNLIR-UHFFFAOYSA-N | |
66951 | |
diethyl 2-oxobutanedioate |
C8H12O5 | |
MFCD00059229 | |
Ethyl Oxalacetate, Oxalacetic Acid Diethyl Ester | |
CHEBI:86608 | |
CCOC(=O)CC(=O)C(=O)OCC |
Safety and Handling
EINECSNumber : (2)-1344
TSCA : No
Recommended Storage : Freezer