missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl Oxalacetate Sodium Salt 95.0+%, TCI America™

Manufacturer:  TCI America O007425G

 View more versions of this product

Catalog No. O007425G



Specifications

Specifications

Diethyl Oxalacetate Sodium Salt
25g
C8H11NaO5
diethyl oxalacetate sodium salt, sodium z-1,4-diethoxy-1,4-dioxo-but-2-en-2-olate, ethyl sodium oxaloacetate, sodium-1,4-diethoxy-1,4-dioxobut-2-en-2-olate, ksc915o5n
CCOC(=O)C=C(C(=O)OCC)[O-].[Na+]
210.161
210.16
≥95.0% (T)
Undesignated
40876-98-0
MFCD00035571
UJZUICGIJODKOS-UHFFFAOYSA-M
sodium;1,4-diethoxy-1,4-dioxobut-2-en-2-olate
44134875
Crystal-Powder at 20°C

Chemical Identifiers

40876-98-0
210.161
UJZUICGIJODKOS-UHFFFAOYSA-M
44134875
CCOC(=O)C=C(C(=O)OCC)[O-].[Na+]
C8H11NaO5
MFCD00035571
diethyl oxalacetate sodium salt, sodium z-1,4-diethoxy-1,4-dioxo-but-2-en-2-olate, ethyl sodium oxaloacetate, sodium-1,4-diethoxy-1,4-dioxobut-2-en-2-olate, ksc915o5n
sodium;1,4-diethoxy-1,4-dioxobut-2-en-2-olate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2624&(2)-2735

RTECSNumber : EK0115500

TSCA : Yes

Documents