missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl n-propylmalonate, 99%, Thermo Scientific Chemicals
$93.51 - $252.21
Chemical Identifiers
CAS | 2163-48-6 |
---|---|
Molecular Formula | C10H18O4 |
Molecular Weight (g/mol) | 202.25 |
MDL Number | MFCD00009168 |
InChI Key | GRRSDGHTSMJICM-UHFFFAOYSA-N |
Synonym | diethyl propylmalonate, diethyl 2-propylmalonate, 1,3-diethyl 2-propylpropanedioate, diethyl n-propylmalonate, diethylpropylmalonate, propylmalonic acid diethyl ester, propanedioic acid, propyl-, diethyl ester, diethyl-n-propylmalonate, malonic acid, propyl-, diethyl ester, diethyl propyl malonate |
PubChem CID | 16552 |
IUPAC Name | diethyl 2-propylpropanedioate |
SMILES | CCCC(C(=O)OCC)C(=O)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2330414
|
Thermo Scientific Chemicals
B2330414 |
25 g |
Each for $93.51
|
|
|||||
AAB2330422
|
Thermo Scientific Chemicals
B2330422 |
100 g |
Each for $252.21
|
|
|||||
Description
Diethyl n-propylmalonate was used in the synthesis of propylacrylic acid (PAA).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl n-propylmalonate was used in the synthesis of propylacrylic acid (PAA).
Solubility
Soluble in Dichloromethane, Ethanol and Ethyl Acetate.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Chemical Identifiers
2163-48-6 | |
202.25 | |
GRRSDGHTSMJICM-UHFFFAOYSA-N | |
16552 | |
CCCC(C(=O)OCC)C(=O)OCC |
C10H18O4 | |
MFCD00009168 | |
diethyl propylmalonate, diethyl 2-propylmalonate, 1,3-diethyl 2-propylpropanedioate, diethyl n-propylmalonate, diethylpropylmalonate, propylmalonic acid diethyl ester, propanedioic acid, propyl-, diethyl ester, diethyl-n-propylmalonate, malonic acid, propyl-, diethyl ester, diethyl propyl malonate | |
diethyl 2-propylpropanedioate |
Specifications
2163-48-6 | |
221°C to 222°C | |
C10H18O4 | |
MFCD00009168 | |
diethyl propylmalonate, diethyl 2-propylmalonate, 1,3-diethyl 2-propylpropanedioate, diethyl n-propylmalonate, diethylpropylmalonate, propylmalonic acid diethyl ester, propanedioic acid, propyl-, diethyl ester, diethyl-n-propylmalonate, malonic acid, propyl-, diethyl ester, diethyl propyl malonate | |
GRRSDGHTSMJICM-UHFFFAOYSA-N | |
diethyl 2-propylpropanedioate | |
16552 | |
99% |
0.987 | |
91°C (197°F) | |
1.4205 | |
25 g | |
Soluble in Dichloromethane,Ethanol and Ethyl Acetate. | |
CCCC(C(=O)OCC)C(=O)OCC | |
202.25 | |
202.25 | |
Diethyl n-propylmalonate |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 218-492-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only