Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl n-butylmalonate, 99%

$41.14 - $148.72

Chemical Identifiers

Chemical Name or Material Diethyl n-butylmalonate
CAS 133-08-4
Molecular Formula C11H20O4
Formula Weight 216.28
MDL Number MFCD00009170
Beilstein 608785
Merck Index 14,1576
Synonym n-Butylmalonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL07539AE

  SDS
alfa aesar™
L07539AE
100mL Each for $41.14

AAL07539AP

  SDS
alfa aesar™
L07539AP
500mL Each for $148.72
Description & Specifications

Specifications

Formula Weight 216.28
Chemical Name or Material Diethyl n-butylmalonate
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Assay 99%
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C11H20O4
Health Hazard 1 Warning
Synonym n-Butylmalonic acid diethyl ester
Beilstein 608785
Quantity 100mL
Merck Index 14,1576
Boiling Point 236° to 239°C
Flash Point 93°C (199°F)
CAS 133-08-4
MDL Number MFCD00009170
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.