Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl methylthiomethylphosphonate, 97%

$90.85 - $360.12

Chemical Identifiers

Chemical Name or Material Diethyl methylthiomethylphosphonate
CAS 28460-01-7
Molecular Formula C6H15O3PS
Formula Weight 198.22
MDL Number MFCD00015131
Beilstein 1705833
Synonym Methylthiomethylphosphonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0057906

  SDS
alfa aesar™
L0057906
5g Each for $90.85

AAL0057914

  SDS
alfa aesar™
L0057914
25g Each for $360.12
Description & Specifications

Specifications

Formula Weight 198.22
Chemical Name or Material Diethyl methylthiomethylphosphonate
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Assay 97%
Molecular Formula C6H15O3PS
MDL Number MFCD00015131
Boiling Point 70° to 72°C (0.2mmHg)
Beilstein 1705833
Flash Point 101°C (213°F)
Synonym Methylthiomethylphosphonic acid diethyl ester
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Quantity 5g
CAS 28460-01-7

Applications
Diethyl methylthiomethylphosphonate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. Diethyl(methylthiomethyl) phosphonate is a reactant for the synthesis of (+)-minfiensine via organocatalytic Diels-Alder/amine cyclization and 6-exo-dig radical cyclization. It is used in the preparation of fused nitrogen, sulfur, and phosphorus containing heterocyclic and relevant difurylpyridazine phosphonates with antimicrobial activity. It is also used as a reactant used in the preparation of fused heterocyclic.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.