missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl methylphosphonite

Manufacturer:  Alfa Aesar™ 3035702

 View more versions of this product

Catalog No. AA3035702



Specifications

Specifications

Diethyl methylphosphonite
1.4168
0.5g
C5H13O2P
UN3279
NSSMTQDEWVTEKN-UHFFFAOYSA-N
diethoxy(methyl)phosphane
85063
Liquid
Strong
Air & Moisture Sensitive
15715-41-0
MFCD00009089
diethyl methylphosphonite, methyldiethoxyphosphine, phosphonous acid, methyl-, diethyl ester, diethoxy methyl phosphane, o,o-diethyl methylphosphonite, diethyl methanephosphonite, phosphonous acid, p-methyl-, diethyl ester, acmc-20akel, diethoxymethylphosphine, diethylmethylphosphonite
CCOP(C)OCC
136.131
136.13

Chemical Identifiers

15715-41-0
136.131
NSSMTQDEWVTEKN-UHFFFAOYSA-N
85063
CCOP(C)OCC
C5H13O2P
MFCD00009089
diethyl methylphosphonite, methyldiethoxyphosphine, phosphonous acid, methyl-, diethyl ester, diethoxy methyl phosphane, o,o-diethyl methylphosphonite, diethyl methanephosphonite, phosphonous acid, p-methyl-, diethyl ester, acmc-20akel, diethoxymethylphosphine, diethylmethylphosphonite
diethoxy(methyl)phosphane
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H226-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see ... on this label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, FLAMMABLE, N.O.S.

TSCA : TSCA

SDS Documents