Learn More
Diethyl methylphosphonite, Thermo Scientific Chemicals
$152.25 - $152.25
Chemical Identifiers
CAS | 15715-41-0 |
---|---|
Molecular Formula | C5H13O2P |
Molecular Weight (g/mol) | 136.13 |
MDL Number | MFCD00009089 |
InChI Key | NSSMTQDEWVTEKN-UHFFFAOYSA-N |
Synonym | diethyl methylphosphonite, methyldiethoxyphosphine, phosphonous acid, methyl-, diethyl ester, diethoxy methyl phosphane, o,o-diethyl methylphosphonite, diethyl methanephosphonite, phosphonous acid, p-methyl-, diethyl ester, acmc-20akel, diethoxymethylphosphine, diethylmethylphosphonite |
PubChem CID | 85063 |
IUPAC Name | diethoxy(methyl)phosphane |
SMILES | CCOP(C)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA3035702
|
Thermo Scientific Chemicals
03035702 |
0.5 g |
Each for $152.25
|
|
|||||
Description
Dimethyl methylphosphonate is a phosphonite reagent used in the generation of phosphinic acid derivative via Arbuzov reaction and as a cyclization reagentin the preparation of 2-substituted penems. It is a chiral intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15715-41-0 | |
136.13 | |
NSSMTQDEWVTEKN-UHFFFAOYSA-N | |
85063 | |
CCOP(C)OCC |
C5H13O2P | |
MFCD00009089 | |
diethyl methylphosphonite, methyldiethoxyphosphine, phosphonous acid, methyl-, diethyl ester, diethoxy methyl phosphane, o,o-diethyl methylphosphonite, diethyl methanephosphonite, phosphonous acid, p-methyl-, diethyl ester, acmc-20akel, diethoxymethylphosphine, diethylmethylphosphonite | |
diethoxy(methyl)phosphane |
Specifications
15715-41-0 | |
C5H13O2P | |
MFCD00009089 | |
UN3279 | |
diethyl methylphosphonite, methyldiethoxyphosphine, phosphonous acid, methyl-, diethyl ester, diethoxy methyl phosphane, o,o-diethyl methylphosphonite, diethyl methanephosphonite, phosphonous acid, p-methyl-, diethyl ester, acmc-20akel, diethoxymethylphosphine, diethylmethylphosphonite | |
NSSMTQDEWVTEKN-UHFFFAOYSA-N | |
diethoxy(methyl)phosphane | |
85063 | |
Liquid |
Strong | |
1.4168 | |
0.5 g | |
Air and moisture sensitive | |
It slowly hydrolyses in water. | |
CCOP(C)OCC | |
136.13 | |
136.13 | |
Diethyl methylphosphonite |
Safety and Handling
GHS H Statement
H300-H310-H330-H226-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
H226-H302
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 239-805-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only