missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl methylmalonate, 99%

Manufacturer:  Alfa Aesar™ A120780B

 View more versions of this product

Catalog No. AAA120780B



Specifications

Specifications

Diethyl methylmalonate
198°C to 199°C
609-08-5
C8H14O4
637028
UPQZOUHVTJNGFK-UHFFFAOYSA-N
diethyl 2-methylpropanedioate
11857
85°C (185°F)
1000g
99%
MFCD00009162
Methylmalonic acid diethyl ester; Methylpropanedioic acid diethyl ester
CCOC(=O)C(C)C(=O)OCC
174.196
174.2

Chemical Identifiers

609-08-5
174.196
UPQZOUHVTJNGFK-UHFFFAOYSA-N
11857
CCOC(=O)C(C)C(=O)OCC
C8H14O4
MFCD00009162
Methylmalonic acid diethyl ester; Methylpropanedioic acid diethyl ester
diethyl 2-methylpropanedioate
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents