missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl Malonate, TCI America™

Manufacturer:  TCI America M002925ML

 View more versions of this product

Catalog No. M002925ML



Specifications

Specifications

Diethyl Malonate
Colorless
25mL
C7H12O4
diethyl malonate, malonic ester, propanedioic acid, diethyl ester, dicarbethoxymethane, malonic acid, diethyl ester, carbethoxyacetic ester, malonic acid diethyl ester, 1,3-diethyl propanedioate, ethyl propanedioate, ethyl malonate van
CCOC(=O)CC(=O)OCC
160.169
160.17
≥99.0% (GC)
199°C
-50°C
105-53-3
MFCD00009195
IYXGSMUGOJNHAZ-UHFFFAOYSA-N
diethyl propanedioate
7761
Clear Liquid at 20°C

Chemical Identifiers

105-53-3
160.169
IYXGSMUGOJNHAZ-UHFFFAOYSA-N
7761
CCOC(=O)CC(=O)OCC
C7H12O4
MFCD00009195
diethyl malonate, malonic ester, propanedioic acid, diethyl ester, dicarbethoxymethane, malonic acid, diethyl ester, carbethoxyacetic ester, malonic acid diethyl ester, 1,3-diethyl propanedioate, ethyl propanedioate, ethyl malonate van
diethyl propanedioate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0913

RTECSNumber : OO0700000

TSCA : Yes

Documents