missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl malonate, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC114450025
Specifications
Diethyl malonate | |
93°C | |
Glass bottle | |
1.055 | |
0.05% max. (K.F.) | |
-50.0°C | |
105-53-3 | |
C7H12O4 | |
02,573 | |
15,3878 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in ethanol, ether and acetone | |
CCOC(=O)CC(=O)OCC | |
160.17 | |
160.17 | |
99+% |
1.0550g/mL | |
Authentic | |
1.4130 to 1.4150 | |
2.14 mPa.s (10°C) | |
199.0°C | |
2.5kg | |
99% min. (GC) | |
C2H5O2CCH2CO2C2H5 | |
10,182; 13,69 | |
1,3-diethyl propanedioate, carbethoxyacetic ester, dicarbethoxymethane, diethyl malonate, ethyl malonate van, ethyl propanedioate, malonic acid diethyl ester, malonic acid, diethyl ester, malonic ester, propanedioic acid, diethyl ester | |
IYXGSMUGOJNHAZ-UHFFFAOYSA-N | |
diethyl propanedioate | |
7761 | |
Liquid |
Chemical Identifiers
105-53-3 | |
160.17 | |
1,3-diethyl propanedioate, carbethoxyacetic ester, dicarbethoxymethane, diethyl malonate, ethyl malonate van, ethyl propanedioate, malonic acid diethyl ester, malonic acid, diethyl ester, malonic ester, propanedioic acid, diethyl ester | |
diethyl propanedioate |
C7H12O4 | |
IYXGSMUGOJNHAZ-UHFFFAOYSA-N | |
7761 | |
CCOC(=O)CC(=O)OCC |
Safety and Handling
EINECSNumber : 203-305-9